HF scientific Powder Pop™ Dispensers for Chlorine Testing
For testing free or total chlorine
Overview
Brand: HF scientific
Manufacturer Part Number: 10506T
Dispenser HF Scientific PPD-2 total chlorine 100
UNSPSC: 60104202
Code: 03
Additional Details:
Additional Details: Weight: 1.00000kg
Product Code. 13161501
Quantity Price
1 £ 30.56 / Each
Estimated Shipment
21-04-2017
Add to basket
Description and Specification
Specification
For Use With (Application) For testing total chlorine
For Use With (Equipment) LaMotte™, Orbeco-Hellige, YSI, palintest, tintometer and Orion photometers
No. of Tests 100.0
Sample Size 10mL
Product Type Dispenser
Documentation