HF scientific Powder Pop™ Dispensers for Chlorine Testing
For testing free or total chlorine
Overview
Brand: HF scientific
Manufacturer Part Number: 10504T
UNSPSC: 60104202
Code: 03
Additional Details:
Additional Details: Weight: 1.00000kg
Product Code. 13131501
Quantity Price
1 £ 31.37 / Each
Call for latest availability
+44 (0)1509 555 500
Add to basket
Description and Specification
Specification
No. of Tests 100.0
Product Type Dispenser
For Use With (Equipment) LaMotte™, Orbeco-Hellige, YSI, palintest, tintometer and Orion photometers
Sample Size 10mL
For Use With (Application) For testing free chlorine
Documentation