missing translation for 'onlineSavingsMsg'
Learn More

Hexyl chloroformate, 98%, Thermo Scientific Chemicals

Catalog Number p-3778754
53.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
6092-54-2
C7H13ClO2
164.63
MFCD00000651
KIWBRXCOTCXSSZ-UHFFFAOYSA-N
hexyl chloroformate, carbonochloridic acid, hexyl ester, n-hexyl chloroformate, chloroformic acid n-hexyl ester, hexyl chlorocarbonate, unii-mu6004c2z3, chloro hexyloxy methanone, chloroformic acid hexyl ester, hexylchloroformate, hexyl=chloroformate
22466
hexyl carbonochloridate
CCCCCCOC(Cl)=O
6092-54-2
C7H13ClO2
164.63
MFCD00000651
KIWBRXCOTCXSSZ-UHFFFAOYSA-N
hexyl chloroformate, carbonochloridic acid, hexyl ester, n-hexyl chloroformate, chloroformic acid n-hexyl ester, hexyl chlorocarbonate, unii-mu6004c2z3, chloro hexyloxy methanone, chloroformic acid hexyl ester, hexylchloroformate, hexyl=chloroformate
22466
hexyl carbonochloridate
CCCCCCOC(Cl)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY
Color Colorless
Density 1.0070g/mL
Boiling Point 60°C to 61°C (7.0 mmHg)
Flash Point 62°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula ClCO2(CH2)5CH3
Refractive Index 1.4230 to 1.4250 (20°C, 589nm)
Quantity 5 g
Specific Gravity 1.007
Formula Weight 164.63
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Hexyl chloroformate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Hexyl chloroformate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'