missing translation for 'onlineSavingsMsg'
Learn More

Hexavalent Chromium, standard solution, Specpure™ Cr|+6 1000μg/mL, Thermo Scientific Chemicals

Catalog Number p-7051910
Click to view available options
Quantity:
100 mL
500 mL
This item is not returnable. View return policy
MFCD00717544
This item is not returnable. View return policy
MFCD00717544

Hexavalent Chromium, standard solution is used in ion chromatography for measurement and determination of soluble Cr(VI).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 100 mL
Color Yellow
Grade Specpure™
Odor Odorless
Solubility Miscible with water
Physical Form Liquid
Chemical Name or Material Hexavalent Chromium standard solution, Cr+6 1000μg/mL

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'