missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexanes, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
237.50 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
1 L
4 L
Packaging:
Aluminum Bottle
Specifications
Formula Weight | 86.18 |
Grade | Meets ACS reagent chemical requirements |
Quantity | 1 L |
Flash Point | −23.00°C |
Packaging | Aluminum Bottle |
Specific Gravity | 1L = 0.66kg |
Chemical Name or Material | Hexanes |
Safety and Handling
Product Identifier
- n-hexane
- Danger
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Aspiration toxicity Category 1
- LONG-TERM AQUATIC HAZARD Chronic 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H315-Causes skin irritation.
- H361f-Suspected of damaging fertility.
- H336-May cause drowsiness or dizziness.
- H373-May cause damage to organs.
- H304-May be fatal if swallowed and enters airways.
- H411-Toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'