missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexamine, Certified AR for Analysis, Fisher Chemical™
106.10 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
Description
Hexamine, Certified AR for Analysis
Specifications
Color | White |
pH | 8.5 to 9.5 |
Packaging | Plastic powder jar |
Quantity | 500 g |
Formula Weight | 140.19g/mol |
Physical Form | Solid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'