missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexamine, Certified AR for Analysis, Fisher Chemical™
207.32 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
Description
Hexamine, Certified AR for Analysis
Specifications
Color | White |
pH | 8.5 to 9.5 |
Packaging | Plastic powder jar |
Quantity | 1 kg |
Formula Weight | 140.19g/mol |
Physical Form | Solid |
Safety and Handling
Product Identifier
- Hexamethylenetetramine
- Warning
- Flammable solid Category 2
- Respiratory sensitiser Category 1
- H317-May cause an allergic skin reaction.
- H228-Flammable solid.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'