missing translation for 'onlineSavingsMsg'
Learn More

Hexamethylphosphorous triamide, 97%, Thermo Scientific Chemicals

Catalog Number p-3778750
Click to view available options
Quantity:
10 mL
50 mL
100 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
1608-26-0
C6H18N3P
163.21
MFCD00008301
XVDBWWRIXBMVJV-UHFFFAOYSA-N
hexamethylphosphorous triamide, tris dimethylamino phosphine, hexametapil, phosphorous triamide, hexamethyl, hexamethyltriamidophosphite, hexamethyltriaminophosphine, phosphine, tris dimethylamino, bis dimethylamino phosphanyl dimethylamine, hmpt van, hexamethyl phosphorus triamide
15355
N-[bis(dimethylamino)phosphanyl]-N-methylmethanamine
CN(C)P(N(C)C)N(C)C
This item is not returnable. View return policy
1608-26-0
C6H18N3P
163.21
MFCD00008301
XVDBWWRIXBMVJV-UHFFFAOYSA-N
hexamethylphosphorous triamide, tris dimethylamino phosphine, hexametapil, phosphorous triamide, hexamethyl, hexamethyltriamidophosphite, hexamethyltriaminophosphine, phosphine, tris dimethylamino, bis dimethylamino phosphanyl dimethylamine, hmpt van, hexamethyl phosphorus triamide
15355
N-[bis(dimethylamino)phosphanyl]-N-methylmethanamine
CN(C)P(N(C)C)N(C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -44°C
Color Colorless to Yellow
Density 0.8900g/mL
Boiling Point 150°C
Flash Point 26°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula [(CH3)2N]3P
Refractive Index 1.4650 to 1.467
Quantity 10 mL
Fieser 01,431; 02,210; 04,247; 06,279; 09,235; 10,199
Specific Gravity 0.89
Solubility Information Solubility in water: reacts
Formula Weight 163.2
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Hexamethylphosphorous triamide
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Hexamethylphosphorous triamide
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'