missing translation for 'onlineSavingsMsg'
Learn More

Hexamethylenetetramine, ACS, 99+%, Thermo Scientific Chemicals

Catalog Number p-7050311
Click to view available options
Quantity:
100 g
500 g
2 kg
100-97-0
C6H12N4
140.19
MFCD00006895
VKYKSIONXSXAKP-UHFFFAOYSA-N
methenamine, hexamethylenetetramine, hexamine, urotropine, 1,3,5,7-tetraazaadamantane, hexamethylenamine, aminoform, methenamin, urotropin, formamine
4101
CHEBI:6824
C1N2CN3CN1CN(C2)C3
100-97-0
C6H12N4
140.19
MFCD00006895
VKYKSIONXSXAKP-UHFFFAOYSA-N
methenamine, hexamethylenetetramine, hexamine, urotropine, 1,3,5,7-tetraazaadamantane, hexamethylenamine, aminoform, methenamin, urotropin, formamine
4101
CHEBI:6824
C1N2CN3CN1CN(C2)C3

Hexamethylenetetramine is used in the manufacturing of phenolic resins liquid preparations and molding compounds. It finds applications in in brake and clutch linings, abrasive products, non-woven textiles and fireproof materials. It is also used as a drug taken by mouth to prevent recurrent urinary tract infections. It is used as a solid fuel in association with 1,3,5-trioxane in campers, hobbyists, the military and relief organizations. It acts as a food additive and as a preservative. It is an important reagent in organic synthesis, especially Duff reaction, Sommelet reaction and in the Delepine reaction. Further, it is used in adhesives, coatings, sealants, and leather preservatives. It is utilized as a formaldehyde releaser for photographic film development and as a hardener for epoxy resins of the bisphenol A type.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 280°C (sublimation)
Color White
Density 1.33 g/mL
Flash Point 250°C (482°F)
Quantity 100 g
UN Number UN1328
Beilstein 2018
Sensitivity Hygroscopic
Merck Index 14,5966
Solubility Information Soluble in water,dimethyl sulfoxide,chloroform,ethanol and acetone. Slightly soluble in ether.
Formula Weight 140.19
Percent Purity ≥99%
Grade ACS
Physical Form Solid
Chemical Name or Material Hexamethylenetetramine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'