missing translation for 'onlineSavingsMsg'
Learn More

Hexamethylbenzene, 98+%, Thermo Scientific Chemicals

Catalog Number p-141994
56.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
87-85-4
C12H18
162.27
MFCD00008523
YUWFEBAXEOLKSG-UHFFFAOYSA-N
hexamethylbenzene, mellitene, benzene, hexamethyl, hexamethylbenzol, hexamethyl benzene, unii-j8sd5741v8, 1,2,3,4,5,6-hexamethyl-benzene, benzene, 1,2,3,4,5,6-hexamethyl, hexamethyl-benzene, hexamethylbenxene;
6908
CHEBI:39001
1,2,3,4,5,6-hexamethylbenzene
CC1=C(C(=C(C(=C1C)C)C)C)C
This item is not returnable. View return policy
87-85-4
C12H18
162.27
MFCD00008523
YUWFEBAXEOLKSG-UHFFFAOYSA-N
hexamethylbenzene, mellitene, benzene, hexamethyl, hexamethylbenzol, hexamethyl benzene, unii-j8sd5741v8, 1,2,3,4,5,6-hexamethyl-benzene, benzene, 1,2,3,4,5,6-hexamethyl, hexamethyl-benzene, hexamethylbenxene;
6908
CHEBI:39001
1,2,3,4,5,6-hexamethylbenzene
CC1=C(C(=C(C(=C1C)C)C)C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 165°C to 168°C
Color White to Yellow
Boiling Point 264°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula C6(CH3)6
Quantity 5 g
Beilstein 05, 450
Fieser 01,426; 01,427; 02,207; 05,323; 06,273; 07,167; 09,234; 13,141; 14,175
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol
Formula Weight 162.27
Percent Purity 98+%
Physical Form Crystalline Powder, Crystals or Needles
Chemical Name or Material Hexamethylbenzene, 99+%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'