missing translation for 'onlineSavingsMsg'
Learn More

Hexafluorophosphoric acid, 60 wt.% solution in water, Thermo Scientific Chemicals

Catalog Number p-141690
Click to view available options
Quantity:
25 g
500 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
16940-81-1, 7664-38-2, 7664-39-3, 13537-32-1, 13779-41-4, 7732-18-5
HF6P
MFCD00011350
Hydrogen hexafluorophosphate
This item is not returnable. View return policy
16940-81-1, 7664-38-2, 7664-39-3, 13537-32-1, 13779-41-4, 7732-18-5
HF6P
MFCD00011350
Hydrogen hexafluorophosphate

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
Linear Formula HPF6
Composition contains the following products of hydrolysis: HPO2F2, H2PO3F, H3PO4 and HF. the presence of these hydrolysis products may rise the strength to over 70%
Fieser 01,42; 02,402
Density 1.7500g/mL
Solubility Information Solubility in water: hydrolyzes slowly.
Formula Weight 145.97
Percent Purity ≥60% (PF6-content)
Packaging Plastic bottle
Specific Gravity 1.75
Quantity 25 g
Physical Form Fuming Solution
Chemical Name or Material Hexafluorophosphoric acid, 60 wt% Solution in Water
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Hexafluorophosphoric acid, 60 wt.% solution in water
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H310-Fatal in contact with skin.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H330-Fatal if inhaled.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'