missing translation for 'onlineSavingsMsg'
Learn More

Hexadecyltrimethylammonium bromide, 99+%, Thermo Scientific Chemicals

Catalog Number 10246520
15.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
10 g
25 g
100 g
500 g
2.5 kg
10 kg
Packaging:
Glass Bottle
Plastic Bottle
Plastic Drum
This item is not returnable. View return policy
57-09-0
C19H42BrN
364.46
MFCD00011772
LZZYPRNAOMGNLH-UHFFFAOYSA-M
5974
CHEBI:3567
[Br-].CCCCCCCCCCCCCCCC[N+](C)(C)C
This item is not returnable. View return policy
57-09-0
C19H42BrN
364.46
MFCD00011772
LZZYPRNAOMGNLH-UHFFFAOYSA-M
5974
CHEBI:3567
[Br-].CCCCCCCCCCCCCCCC[N+](C)(C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY
Color White
Physical Form Crystalline Powder
Melting Point 230.0°C
Decomposition Information >230°C
Assay Percent Range 99+%
Linear Formula CH3(CH2)15N(CH3)3Br
Quantity 25 g
Beilstein 04, IV, 819
Fieser 15,77
Merck Index 15, 2024
Synonym Cetrimonium Bromide, Cetyltrimethylammonium Bromide, CTABr
Solubility Information Solubility in water: 13g/L (20°C). Other solubilities: soluble in methanol (>200g/L at 20 °C), ethanol, insoluble in benzene, diethyl ether, chloroform, sparingly soluble in acetone
Flash Point 244°C
IUPAC Name hexadecyltrimethylazanium bromide
Formula Weight 364.45
Infrared Spectrum Authentic
Percent Purity ≥99%
Packaging Plastic Bottle
Chemical Name or Material Hexadecyltrimethylammonium Bromide
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Hexadecyltrimethylammonium bromide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H315-Causes skin irritation.
  • H373-May cause damage to organs.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'