missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexachloroplatinic(IV) Acid Hydrate, Extra Pure, SLR, Fisher Chemical™
212.15 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
1 g
5 g
Specifications
Melting Point | 60°C |
Color | Amber |
Packaging | Vial |
Quantity | 1 g |
Formula Weight | 409.82g/mol |
Physical Form | Powder |
Safety and Handling
Product Identifier
- Hexachloroplatinic(IV) acid hydrate
- Danger
- Skin corrosion/irritation Category 1 B
- Acute toxicity Category 3
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- H314-Causes severe skin burns and eye damage.
- H301-Toxic if swallowed.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H317-May cause an allergic skin reaction.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Hydrogen hexachloroplatinate(IV)hydrate,Chloroplatinic acid
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'