missing translation for 'onlineSavingsMsg'
Learn More

Hexacarbonyltungsten, 97%, Thermo Scientific Chemicals

Catalog Number p-4904542
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
14040-11-0
C6O6W
MFCD00011462
Tungsten hexacarbonyl
This item is not returnable. View return policy
14040-11-0
C6O6W
MFCD00011462
Tungsten hexacarbonyl

Hexacarbonyltungsten is used as a precursor to a Fischer carbine complex, which used in the hiyama cross-coupling reactions. It is widely used in electron beam-induced deposition technique as well as a source of tungsten atoms. It is utilized to desulfurize organosulfur compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 172°C to 174°C
Color White
Density 2.65
Assay Percent Range 97%
Odor Nearly odorless
Quantity 10 g
Solubility Information Slightly soluble in tetrahydrofuran. Insoluble in water.
Formula Weight 351.91
Vapor Pressure 1.596hPa (1mm Hg) at 67°C
Physical Form Crystalline
Chemical Name or Material Hexacarbonyltungsten
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'