missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Hexaamminenickel(II) iodide, 99.999% (metals basis)

Brand:  Alfa Aesar™ 044481.14

Code : NEW

Additional Details : CAS Number : 13859-68-2 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 11480820

  • £106.40 / 25g


EU Stock 1
Add to basket

Specifications

Chemical Identifiers

Hexaamminenickel (II) iodide
Ni(NH3)6I2
MFCD00058808
13859-68-2
414.68

Specifications

Hexaamminenickel (II) iodide
13859-68-2
99.999%
Ni(NH3)6I2
MFCD00058808
25g
Crystalline
(metals basis)
414.68
Videos
SDS