Learn More
HERC1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA562033
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: KGKLLSSQEG MISFDCFMTI LMQMRRSLGS SADRSQWREP TRTSDGLCSL YEAALCLFEE VCRMASDYSR TCASPDSIQT GDAPIVSETC EVYVWGSNSS HQLVEGTQEK ILQPK Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.
HERC1 Polyclonal antibody specifically detects HERC1 in Human samples. It is validated for Immunohistochemistry.Specifications
HERC1 | |
Polyclonal | |
Unconjugated | |
HERC1 | |
guanine nucleotide exchange factor p532; hect (homologous to the E6-AP (UBE3A) carboxyl terminus) domain and RCC1 (CHC1)-like domain (RLD) 1; HECT and RLD domain containing E3 ubiquitin protein ligase family member 1; HECT domain and RCC1-like domain-containing protein 1; HECT-type E3 ubiquitin transferase HERC1; HERC1; p532; p619; Probable E3 ubiquitin-protein ligase HERC1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8925 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q15751 | |
HERC1 | |
Recombinant protein corresponding to Human HERC1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only