Learn More
Thermo Scientific™ Heracell™ Gas Tight Inner Door
Material: Glass; For Use With: Heracell VIOS 250i CO2 Incubator
Brand:  Thermo Scientific™ 51901127
Additional Details : Weight : 0.10000kg
Product Code. 15582720
  • £3312.88 / Each
Estimated Shipment: 18-07-2024
to see stock.
Add to basket
Description
Description
6 gas tight doors replaces single inner door configuration
Specifications
Specifications
Gsa tight interior glass doors
LED
Inner Door
Heracell VIOS 250i CO2 incubator
Glass
6 doors