Learn More
O2 Control for Heracell™ VIOS 160i and Forma™ Steri-Cycle™ i160 Incubators
Transform your Heracell™ VIOS 160i and Forma™ Steri-Cycle i160 Incubators into a variable control oxygen incubator with O2 control.
Brand:  Thermo Scientific Heraeus 51901146
UNSPSC : 41100000
Code : NEW
Additional Details : Weight : 5.00000kg
 View more versions of this product
Product Code. 15353012
  • £3645.00 / Each
Call for latest availability
+44 (0)1509 555 500
Add to basket
Specifications Description & Specifications
Specifications
5 to 90% O2 control with 3 door inner gas tight screen door
Heracell™ VIOS 160i and Forma™ Steri-Cycle™ i160 Incubators