Learn More
Thermo Scientific™ Heracell™ VIOS™ 160i CO2 Incubator with Cell Locker™ System
Protected chambers for your most sensitive cells.  A breakthrough in cell culture management that maximizes your sample security
£11277.03 - £12370.09
Products 2
Product Code Brand Description CO2 Sensor Technology Electrical Requirements Price Quantity & Availability  
Product Code Brand Description CO2 Sensor Technology Electrical Requirements Price Quantity & Availability  
15814295 Thermo Scientific™
51032125
Single 165L stainless steel chamber configured for Cell Locker system, (Cell Locker chambers ordered separately) IR180SI 230 V, 50/60 Hz £11277.03
Each View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
15824295 Thermo Scientific™
51032129
Single 165L copper chamber configured for Cell Locker system, (Cell Locker chambers ordered separately) IR180SI 230 V, 50/60 Hz £12370.09
Each View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
Description
Description
CO2 Incubator with Cell Locker System
  • 165L interior chamber with the new 6 segmented, gas ight inner door configuration and 3 shelves designed to accommodate 6 individual Cell Locker chambers.
  • Choice of electropolished stainless steel or 100% pure copper chamber interior.
  • Temperature resistant, bulb-free IR CO2 sensor (IR180Si).
  • THRIVE airflow technology provides enhanced stability,tight uniformity, and fast recovery.
  • ISO Class 5 HEPA filtration provides clean room air quality in the chamber.
  • Thermo Scientific iCAN touchscreen interface provides bright at-glance monitoring and complete data visibility to monitor all incubator interactions and parameters.
  • Thermo Scientific Steri-Run on-demand, overnight 180˚C sterilization cycle achieves 12 log Sterility Assurance Level (SAL) total sterilization of chamber (excluding Cell Locker chambers )
Stability of Conditions
  • Cultures spend more time in conditions that best mimic the in vivo state.
  • When one Cell Locker chamber is opened, the other Cell Locker chambers maintain stable conditions, preserving the in vitro environment.
  • Conditions remain stable within these spatial deviations - temperature +/-0.3˚C , humidity +/-3%, and CO2 +/-0.2%.
  • For consistent results, the uniformity in each Cell Locker chamber is +/-0.3˚C
  • Compared to the standard single glass door design, the temperature drop in the opened Cell Locker chamber is reduced by 50%.
  • With the Cell Locker system, gas consumption from a door opening is reduced by 50%, saving time and money.
Sample Protection
  • Individual chambers maximize security against costly cross contamination.
  • Each individual Cell Locker chamber is effectively a quarantine chamber.
  • Dual 0.2μm membrane filters permits air circulation but exclude microbial contaminants.
  • The replacable 0.2μm membrane filter has an effective pore size of 0.02μm when filtering air and is hydrophobic, oleophobic, resistant to organic solvents and tested for biosafety and low cytotoxicity.
  • Independent 3rd party tests validate protection against circulating microorganisms (when used as directed)
Flexibility of Use
  • The Cell Locker System can be configured to fit unique needs of your lab.
  • Different cell types, samples, users or projects can be segregated within shared incubator space.
  • Each Cell Locker chamber holds 9 each T-75 cell culture flasks, 20 each 6-well plates, or 24 each 96-well plates.
  • Dishes and flasks can be removed individually on a work tray or inside the entire Cell Locker chamber with optional transport cover, thus protecting samples from outside air.
  • The non skid tray slides out to allow access to culture vessels, or the entire tray can be removed.
  • The Cell Locker chamber can be manually disinfected or can be autoclaved.
Order Info
Please order Cell Lockers separately if not included. 50151650 is a single Cell Locker with membrane filters with stainless steel work tray and transport cover.; 50154739 is a single Cell Locker with membrane filters with copper work tray and transport cover