missing translation for 'onlineSavingsMsg'
Learn More

HER-2/c-erbB-2/neu Ab-8 Mouse Monoclonal Antibody, Epredia™

Recommended for Immunohistochemistry, Western Blotting and Immunoprecipitation (Native and Denatured), Epredia™ HER-2/c-erbB-2/neu Ab-8, Mouse Monoclonal Antibody provides accurate, reproducible results.

£120.00 - £515.00

Specifications

Antigen HER-2/c-erbB-2/neu Ab-8
Clone e2-4001
Applications Immunoprecipitation
Classification Monoclonal
Form Concentrated
View More Specs

Products 7
Product Code Brand Form Conjugate Concentration Quantity Price Quantity & Availability  
Product Code Brand Form Conjugate Concentration Quantity Price Quantity & Availability  
12661017
View Documents Promotion Details
Epredia
MS-325-B0
Concentrated Biotin 200μg/mL 100 μL
£162.00
100µL
View alternative products
Save Up to 
»
Estimated Shipment: 03-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
12691017
View Documents Promotion Details
Epredia
MS-325-P0
Concentrated Unconjugated 200μg/mL 100 μL
£120.00
100µL
View alternative products
Save Up to 
»
Estimated Shipment: 03-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
12671017
View Documents Promotion Details
Epredia
MS-325-B1
Concentrated Biotin 200μg/mL 500 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12601027
View Documents Promotion Details
Epredia
MS-325-P1
Concentrated Unconjugated 200μg/mL 500 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12681017
View Documents Promotion Details
Epredia
MS-325-P
Concentrated Unconjugated 200μg/mL 1 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12611027
View Documents Promotion Details
Epredia
MS-325-P1ABX
Concentrated Unconjugated 1mg/mL 100 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12621027
View Documents Promotion Details
Epredia
MS-325-PABX
Concentrated Unconjugated 1mg/mL 200 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

c-erbB-2 is a receptor tyrosine kinase. It exhibits extracellular domains with two cysteine-rich sequences, and a cytoplasmic tyrosine kinase domain flanked by large hydrophilic tails that carry several tyrosine autophosphorylation sites. Approximately 25% of primary breast and ovarian tumors were found to overexpress the protein. The c-erbB-2 protein is overexpressed in a variety of carcinomas especially those of breast and ovary.

Host Species: Mouse

Clone: e2-4001

Isotype: IgG1

Species Reactivity: Human, Mouse and Rat. Others not tested.

Epitope: Cytoplasmic domain

Immunogen: Cytoplasmic domain of recombinant human HER-2/c-erbB-2 oncoprotein

Molecular Weight: 185 kDa

Positive Control: SKBR-3 cells, or T47D cells, or Breast CA

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry
  • Western Blotting
  • Immunoprecipitation (Native and Denatured)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

HER-2/c-erbB-2/neu Ab-8
Immunoprecipitation
Concentrated
RUO
Human, Mouse, Rat
IgG1
e2-4001
Monoclonal
Mouse
Oncogenesis
Cytoplasmic domain of recombinant human c-erbB-2/HER-2 oncoprotein
Primary
Videos
SDS
Documents

Documents

Product Certifications