missing translation for 'onlineSavingsMsg'
Learn More

Heptanophenone, 98+%, Thermo Scientific™

Catalog Number p-140513
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1671-75-6
C13H18O
190.29
MFCD00009539
UXMQORVHJMUQFD-UHFFFAOYSA-N
heptanophenone, 1-heptanone, 1-phenyl, n-heptanophenone, unii-kx07wp06jy, kx07wp06jy, enanthophenone, hexyl phenyl ketone, 1-phenyl-heptan-1-one, 1-phenyl-1-heptanone #
74282
1-phenylheptan-1-one
CCCCCCC(=O)C1=CC=CC=C1
This item is not returnable. View return policy
1671-75-6
C13H18O
190.29
MFCD00009539
UXMQORVHJMUQFD-UHFFFAOYSA-N
heptanophenone, 1-heptanone, 1-phenyl, n-heptanophenone, unii-kx07wp06jy, kx07wp06jy, enanthophenone, hexyl phenyl ketone, 1-phenyl-heptan-1-one, 1-phenyl-1-heptanone #
74282
1-phenylheptan-1-one
CCCCCCC(=O)C1=CC=CC=C1
Melting Point 17°C
Color Yellow
Density 0.9400g/mL
Boiling Point 155°C (15.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula C6H5CO(CH2)5CH3
Refractive Index 1.5050 to 1.508
Quantity 5 g
Specific Gravity 0.94
Formula Weight 190.29
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material Heptanophenone, 98+%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'