missing translation for 'onlineSavingsMsg'
Learn More

Heptanoic acid, 98%, Thermo Scientific Chemicals

Catalog Number 10783351
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
111-14-8
C7H14O2
130.19
MFCD00004426
MNWFXJYAOYHMED-UHFFFAOYSA-N
enanthic acid, oenanthic acid, n-heptanoic acid, enanthylic acid, heptoic acid, n-heptoic acid, n-heptylic acid, heptylic acid, oenanthylic acid, 1-hexanecarboxylic acid
8094
CHEBI:45571
heptanoic acid
CCCCCCC(=O)O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
111-14-8
C7H14O2
130.19
MFCD00004426
MNWFXJYAOYHMED-UHFFFAOYSA-N
enanthic acid, oenanthic acid, n-heptanoic acid, enanthylic acid, heptoic acid, n-heptoic acid, n-heptylic acid, heptylic acid, oenanthylic acid, 1-hexanecarboxylic acid
8094
CHEBI:45571
heptanoic acid
CCCCCCC(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -10.5°C
Color White
Density 0.9100g/mL
Boiling Point 223.0°C to 223.4°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3(CH2)5CO2H
Refractive Index 1.4210 to 1.4240
Quantity 250 mL
Beilstein 02,338
Merck Index 15,4695
Specific Gravity 0.91
Solubility Information Solubility in water: 0.24g/100mL (15°C). Other solubilities: soluble in alcohol, ether, dmf, chloroform, petrol, soluble in most organic solvents
Viscosity 3.40 mPa.s (30°C)
Formula Weight 130.19
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Heptanoic acid, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Heptanoic acid
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'