missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heptane, puriss., Honeywell™
Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
25 L
Unit Size:
25 L
Specifications
Specifications
| CAS | 142-82-5 |
| Density | 0.684g/mL at 25°C |
| Quantity | 25 L |
| Explosion Limit | 0.07 |
| Flash Point | −4°C (24.8°F) |
| Ignition Point | 433°F |
| Molecular Formula | C7H16 |
| Linear Formula | CH3(CH2)5CH3 |
| MDL Number | MFCD00009544 |
| Vapor Pressure | 83mmHg (37.7°C), 40mmHg (20°C) |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction