missing translation for 'onlineSavingsMsg'
Learn More

Heptakis-6-iodo-6-deoxy-β-cyclodextrin, Thermo Scientific™

Catalog Number p-140474
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
This item is not returnable. View return policy
30754-23-5
C42H63I7O28
1904.28
MFCD00803290
YODKQJPYQJRHEQ-FOUAGVGXSA-N
heptakis-6-iodo-6-deoxy-beta-cyclodextrin
11105283
C(C1C2C(C(C(O1)OC3C(OC(C(C3O)O)OC4C(OC(C(C4O)O)OC5C(OC(C(C5O)O)OC6C(OC(C(C6O)O)OC7C(OC(C(C7O)O)OC8C(OC(O2)C(C8O)O)CI)CI)CI)CI)CI)CI)O)O)I
This item is not returnable. View return policy
30754-23-5
C42H63I7O28
1904.28
MFCD00803290
YODKQJPYQJRHEQ-FOUAGVGXSA-N
heptakis-6-iodo-6-deoxy-beta-cyclodextrin
11105283
C(C1C2C(C(C(O1)OC3C(OC(C(C3O)O)OC4C(OC(C(C4O)O)OC5C(OC(C(C5O)O)OC6C(OC(C(C6O)O)OC7C(OC(C(C7O)O)OC8C(OC(O2)C(C8O)O)CI)CI)CI)CI)CI)CI)O)O)I
Chelation/Complexation reagent
TRUSTED_SUSTAINABILITY
Melting Point 218°C to 222°C
Color White
Packaging Glass bottle
Quantity 100 mg
Solubility Information Solubility in water: .. Other solubilities: soluble in dmf,dmso,hot methylethyleneglycol
Formula Weight 1904.28
Physical Form Solid
Chemical Name or Material Heptakis-6-iodo-6-deoxy-β-cyclodextrin

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'