missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals HEPES sodium salt, 99%
Click to view available options
Quantity:
25 g
100 g
Description
HEPES [N-(2-hydroxyethyl)piperazine-N'-(2-ethanesulfonic acid)] is a general purpose buffer for biological research. Its applications include, tissue culture, Phosphorylation and photophosphorylation, fixative Transmission electron microscopy, protein synthesis, preventing binding to non-receptor materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
HEPES [N-(2-hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid)] is a general purpose buffer for biological research. Its applications include, tissue culture, Phosphorylation and photophosphorylation, fixative Transmission electron microscopy, protein synthesis, preventing binding to non-receptor materials.
Solubility
It is soluble in water.
Notes
Stable under ordinary conditions. Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
HEPES [N-(2-hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid)] is a general purpose buffer for biological research. Its applications include, tissue culture, Phosphorylation and photophosphorylation, fixative Transmission electron microscopy, protein synthesis, preventing binding to non-receptor materials.
Solubility
It is soluble in water.
Notes
Stable under ordinary conditions. Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Specifications
Quantity | 25 g |
Sensitivity | Hygroscopic |
Solubility Information | It is soluble in water. |
IUPAC Name | sodium;2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonate |
Formula Weight | 260.28 |
Percent Purity | 99% |
Chemical Name or Material | HEPES sodium salt |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'