missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HEPES, Free Acid, Ultrapure Bioreagent, J.T. Baker™
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly bottle
Description
- For Liquid Chromatography and Molecular Biology applications; For Laboratory,Research,or Manufacturing Use
Specifications
Quantity | 500 g |
Packaging | Poly bottle |
Specific Gravity | 1L = 0.00kg |
Formula Weight | 238.31 |
Grade | Ultrapure Bioreagent |
Chemical Name or Material | HEPES, Free Acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'