Learn More
Thermo Scientific Chemicals HEPES-buffered saline (2X, pH 7.0), low endotoxin
Brand: Thermo Scientific Chemicals J67505.AK
59.40 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.00025kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
HEPES-buffered saline (2X, pH 7.0) | |
Colorless | |
250 mL |
low endotoxin | |
Liquid |
RUO – Research Use Only