missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Hepassocin/FGL1 Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 1 publication
POA
Specifications
Antigen | Hepassocin/FGL1 |
---|---|
Clone | 198401 |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Classification | Monoclonal |
Description
Hepassocin/FGL1 Monoclonal specifically detects Hepassocin/FGL1 in Human samples. It is validated for Western Blot.Specifications
Hepassocin/FGL1 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
FGL1, fibrinogen-like 1, fibrinogen-like protein 1, Fibrinogen-like-protein 1, FREP1, Hepassocin, hepatocellular carcinoma-related sequence, Hepatocyte-derived fibrinogen-related protein 1, HFREP-1, HFREP1MGC12455, HP-041, LFIRE1, LFIRE-1, Liver fibrinogen-related protein 1 | |
FGL1 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
198401 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
BAB70690 | |
2267 | |
Mouse myeloma cell line NS0-derived recombinant human Hepassocin Met1-Ile312 Accession # BAB70690 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Hepassocin in direct ELISAs and Western blots. In direct ELISAs, weak cross-reactivity with rat Hepassocin is observed. In Western blots, 100% cross-reactivity with rat Hepassocin is observed. |
For Research Use Only