missing translation for 'onlineSavingsMsg'
Learn More

Heparin, Fisher BioReagents

Brand:  Fisher Bioreagents BP2524-100

Code : 32

Additional Details : CAS Number : 9041-08-1 Weight : 0.05010kg

Quantity 100mg
Packaging Poly Vial

 View more versions of this product

Product Code. 10639163

  • £79.62 / 100mg


In Stock 11
EU Stock 29
Add to basket

Specifications

Chemical Identifiers

Heparin
From Porcine Intestinal Mucosa
9041-08-1
≈5000

Specifications

Heparin
100mg
Report U/mg (Anti-XA)
9041-08-1
≤0.03 EU/HEP
0.003% max.
1.3 to 2.5%
≥140hep/mg (dry basis)
≈5000
White to Off-white
5.0 to 7.5
From Porcine Intestinal Mucosa
Vapor Pressure: Negligible
Pass Test
≤5.0%
Poly Vial
Powder/Solid
15, 4688
Videos
SDS Documentation

Technical Documents