missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Heparan Sulfate 6-O-Sulfotransferase 2/HS6ST2 Antibody, R&D Systems™
Goat Polyclonal Antibody
POA
Specifications
Antigen | Heparan Sulfate 6-O-Sulfotransferase 2/HS6ST2 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL |
Applications | Western Blot, Immunoprecipitation |
Classification | Polyclonal |
Description
Heparan Sulfate 6-O-Sulfotransferase 2/HS6ST2 Polyclonal specifically detects Heparan Sulfate 6-O-Sulfotransferase 2/HS6ST2 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Heparan Sulfate 6-O-Sulfotransferase 2/HS6ST2 | |
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
Polyclonal | |
Goat | |
Human | |
EC-1, heparan sulfate 6-O-sulfotransferase 2 | |
HS6ST2 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
RUO | |
Q96MM7 | |
90161 | |
Mouse myeloma cell line NS0-derived recombinant human Heparan Sulfate 6-O-Sulfotransferase 2/HS6ST2 Gln179-Arg605 Accession # Q96MM7 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human Heparan Sulfate 6-O-Sulfotransferase 2/HS6ST2 in direct ELISAs and Western blots. |
For Research Use Only