Learn More
Human Hematopoietic Lineage Antibody Cocktail, APC, eBioscience™, Invitrogen™
Antibody Cocktail
Brand: Affymetrix eBioscience 22-7776-72
Code : NEW
Additional Details : Weight : 0.23750kg
Description
Description: The Human Hematopoietic Lineage APC Cocktail contains APC-conjugated antibodies that can be used to identify, enrich and/or deplete cells committed to the T, B, NK, myeloid and erythroid lineages based on their expression of cell surface antigens. This cocktail contains the following antibodies: Anti-Human CD2 (RPA-2.10) APC Anti-Human CD3 (OKT3) APC Anti-Human CD14 (61D3) APC Anti-Human CD16 (CB16) APC Anti-Human CD19 (HIB19) APC Anti-Human CD56 (TULY56) APC Anti-Human CD235a (HIR2) APC Applications Reported: The Human Hematopoietic Lineage APC Cocktail has been reported for use in flow cytometric analysis. Applications Tested: The Human Hematopoietic Lineage APC Cocktail has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 20 μL per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
Human Hematopoietic Lineage Cocktail antibody specifically detects Human Hematopoietic Lineage in Human samples. It is validated for Flow Cytometry.Specifications
Human Hematopoietic Lineage | |
Cocktail | |
20 μL/Test | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
Mouse | |
50 Tests | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
RPA-2.10, OKT3, 61D3, CB16, HIB19, TULY56, HIR2, OKT3, 61D3, CB16, HIB19, TULY56, HIR2 | |
APC | |
17A2; Hematopoietic Lineage Cocktail; M1/70; RA3-6B2; RB6-8C5; TER-119 | |
Affinity chromatography | |
RUO | |
Human | |
Antibody |
For Research Use Only.