Learn More
Scientific Cell Company Hellma Analytics Liquid Filters
Brand:  Scientific Cell Company 667005
Code : NEW
Additional Details : Weight : 0.25000kg
 View more versions of this product
Product Code. 10165983
Quantity Price
1 £569.00 / Each
5 £521.15 / Each
Estimated Shipment: 25-11-2021
Specifications
DIN EN ISO 17025, NIST
For testing the wavelength accuracy
Liquid filter
241.15, 287.15, 361.5, 536.3, 640.5nm
Holmium oxide in perchloric acid