missing translation for 'onlineSavingsMsg'
Learn More

HELIOS Armenian Hamster anti-Canine, Human, Mouse, PE-Cyanine7, Clone: 22F6, eBioscience™

Armenian Hamster Monoclonal Antibody

£94.25 - £232.00

Specifications

Antigen Helios
Clone 22F6
Target Species Canine, Human, Mouse
Conjugate PE-Cyanine7
Isotype IgG
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15568686
View Documents
Affymetrix eBioscience
25-9883-41
25 Tests
£94.25
25 tests
View alternative products
Save Up to 
»
Estimated Shipment
04-01-2021
Please to purchase this item. Need a web account? Register with us today!
15578686
View Documents
Affymetrix eBioscience
25-9883-42
100 Tests
£232.00
100 tests
View alternative products
Save Up to 
»
In stock, ready to ship: 9 Please to purchase this item. Need a web account? Register with us today!
Description

Description

A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 µL of cell sample + 100 µL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 488-561 nm; Emission: 775 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered. This gene encodes a member of the Ikaros family of zinc-finger proteins. Three members of this protein family (Ikaros, Aiolos and Helios) are hematopoietic-specific transcription factors involved in the regulation of lymphocyte development. This protein forms homo- or hetero-dimers with other Ikaros family members, and is thought to function predomitly in early hematopoietic development. Multiple transcript variants encoding different isoforms have been found for this gene, but the biological validity of some variants has not been determined.
Specifications

Specifications

Helios
Canine, Human, Mouse
IgG
Monoclonal
Liquid
Antibody
4° C, store in dark, DO NOT FREEZE!
22F6
PE-Cyanine7
Flow Cytometry
Primary
Monoclonal
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2
RUO
SDS

For Research Use Only.