missing translation for 'onlineSavingsMsg'
Learn More

HELIOS, PE, clone: 22F6, eBioscience™

Armenian Hamster Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-9883-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15547266

  • POA / 25 tests

This item cannot be ordered online because the price is subject to change. Please contact our Product Support team on 01509 555500 for assistance.


Description

Description

Description: This 22F6 monoclonal antibody reacts with human, mouse and canine Helios. Helios is a zinc-finger transcription factor first identified for its similarity to Ikaros. It is expressed in hematopoietic stem cells in both the embryo and adult mice. While the highest expression is found in regulatory T cells, Helios is not expressed in mature B cells, dendritic cells or myeloid cells. Helios can form a homodimer as well as heterodimerize with other Ikaros family members to regulate transcription. Eight isoforms of Helios exist, some of which possess a dominant-negative function that has also been found to be upregulated in lymphoid malignancies, including T-acute lymphoblastic leukemia.22F6 has been reported to cross react with rat Helios.Applications Reported: This 22F6 antibody has been reported for use in intracellular staining followed by flow cytometric analysis.Applications Tested: This 22F6 antibody has been pre-titrated and tested by intracellular staining and flow cytometric analysis of mouse lymph node cell using the Foxp3/Transcription Factor Buffer Set (cat. 00-5523) and protocol. Please see Best Protocols Section (Staining intracellular Antigens for Flow Cytometry) for staining protocol (refer to Protocol B: One-step protocol for intracellular (nuclear) proteins). This can be used at 5 μL (0.03 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Helios
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P81183, Q9UKS7
HELIOS, ZNF1A2, ZNFN1A2, ikaros family zinc finger protein 2, zinc finger DNA binding protein Helios, zinc finger protein Helios, zinc finger protein, subfamily 1A, 2 (Helios)
Armenian Hamster
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
22F6
PE
IKZF2
Conjugated
IKZF2
IgG
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
22807
Videos

For Research Use Only.