missing translation for 'onlineSavingsMsg'
Learn More

HEAT hydrochloride, Tocris Bioscience™

Highly selective α1 antagonist

Brand:  Tocris (Bio-Techne) 053510

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13447693

  • £148.00 / 10mg

Estimated Shipment
17-09-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Very selective α1-adrenoceptor antagonist, precursor of the 3-[125I]-derivative.
Specifications

Chemical Identifiers

30007-39-7
331.84
heat hydrochloride, heat, 2-4-hydroxyphenethyl amino methyl-3,4-dihydronaphthalen-1 2h-one hydrochloride, 2-?-4-hydroxyphenyl ethyl aminomethyl-1-tetralone hydrochloride, 2-4-hydroxyphenethylamino methyl-3,4-dihydronaphthalen-1 2h-one hydrochloride, 2-2-4-hydroxyphenyl ethylamino methyl-3,4-dihydro-2h-naphthalen-1-one hydrochloride, 2-2-4-hydroxyphenyl ethyl amino methyl-3,4-dihydro-2h-naphthalen-1-one hydrochloride
2-[[2-(4-hydroxyphenyl)ethylamino]methyl]-3,4-dihydro-2H-naphthalen-1-one;hydrochloride
C19H22ClNO2
VCZXZECZIRGUCZ-UHFFFAOYSA-N
6917680
C1CC2=CC=CC=C2C(=O)C1CNCCC3=CC=C(C=C3)O.Cl

Specifications

HEAT hydrochloride
30007-39-7
heat hydrochloride, heat, 2-4-hydroxyphenethyl amino methyl-3,4-dihydronaphthalen-1 2h-one hydrochloride, 2-?-4-hydroxyphenyl ethyl aminomethyl-1-tetralone hydrochloride, 2-4-hydroxyphenethylamino methyl-3,4-dihydronaphthalen-1 2h-one hydrochloride, 2-2-4-hydroxyphenyl ethylamino methyl-3,4-dihydro-2h-naphthalen-1-one hydrochloride, 2-2-4-hydroxyphenyl ethyl amino methyl-3,4-dihydro-2h-naphthalen-1-one hydrochloride
C1CC2=CC=CC=C2C(=O)C1CNCCC3=CC=C(C=C3)O.Cl
331.84
331.84
10mg
C19H22ClNO2
VCZXZECZIRGUCZ-UHFFFAOYSA-N
2-[[2-(4-hydroxyphenyl)ethylamino]methyl]-3,4-dihydro-2H-naphthalen-1-one;hydrochloride
6917680
>98%
Videos
Documents