missing translation for 'onlineSavingsMsg'
Learn More

Healthcare Pouch

Brand:  Vernacare AAAMC90230

Code : 03

Additional Details : Weight : 0.50000kg

Product Code. 12739075

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

Specifications

Specifications

Self-seal
2000/Pk.
90 mm
For Use in high vacuum, high temperature steam autoclaves and other sterilization systems
mm
203 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers