missing translation for 'onlineSavingsMsg'
Learn More

Synergy Healthcare Pouch code P9 flat peelable 150mm x 203mm

Brand:  Synergy Healthcare PF150210

Pouch code P9 flat peelable 150mm x 203mm

Code : 03

Additional Details : Weight : 0.20000kg

Product Code. 12739005

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.