missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kartell™ HDPE Standardized Screw Caps
Click to view available options
Closure Size:
GL18
Description
- Made of HDPE (High-density Polyethylene)
- For use with PE bottles DIN 168
- Easy to use screw cap
Specifications
Closure Material | HDPE |
Closure Size | GL 25 |
Closure Type | Screw Cap |
For Use With (Equipment) | Kartell™ Bottles 15841 - 15861 |
Product Type | HDPE Screw Cap |
Color | White |
Material | HDPE |
Screw Material | HDPE |
Product Line | Cap |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'