missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ HDPE General Purpose Funnels

 Manufactured from HDPE for optimum strength and chemical resistance. Kartell™ HDPE General Purpose Funnels provide safety and precision in filling bottles in the laboratory.

Brand:  Kartell™ 4179

UNSPSC : 41121809

Code : A0

Additional Details : Weight : 0.60000kg

 View more versions of this product

Product Code. 10081101

  • £13.70 / Each


In Stock 5
Add to basket

Description

Description

  • Suitable for a wide range of liquids
  • Upstand on rim reduces potential overflow
  • Outer ribbing eliminates back pressure 
  • Hanging eye for storage
Specifications

Specifications

Suitable for a wide range of liquids
57mm
138mm
10000mL
420mm
HDPE