missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ HDPE General Purpose Funnels

Manufactured from HDPE for optimum strength and chemical resistance. Kartell™ HDPE General Purpose Funnels provide safety and precision in filling bottles in the laboratory.

Brand:  Kartell™ 173

Code : 02

Additional Details : Weight : 0.15604kg

 View more versions of this product

Product Code. 12553407

  • POA / Pack of 5

This item cannot be ordered online because the price is subject to change. Please contact our Product Support team on 01509 555500 for assistance.
View alternative products »


You may only order up to the available stock. For more details call customer services on 01509 555 500

Description

Description

  • Suitable for a wide range of liquids
  • Upstand on rim reduces potential overflow
  • Outer ribbing eliminates back pressure 
  • Hanging eye for storage
Specifications

Specifications

20mm
Suitable for a wide range of liquids
450mL
130mm
43mm
HDPE (High-density Polyethylene)
Videos
Documentation

Technical Documents