missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Scienceware™ HDPE Biohazard Bags

Protects against dangerous tears and leaks with extra strong, high impact polypropylene biohazard disposal bags. Bel Art ™ HDPE Biohazard Bags are printed with standard biohazard warning symbol and precautionary procedures in four languages: English, French, German and Spanish.

Brand:  Bel-Art™ Scienceware™ 06494-06

Code : A0

Additional Details : Weight : 0.80000kg

Product Code. 11957165

  • £987.23 / Pack of 200


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

  • Pass ASTM 1709 Dart Impact and ASTM 1922 Tear Resistance Tests
  • Extra heavy-duty; 0.05mm (2 mil) thick blended polymer
  • Autoclavable to 135°C (275°F)
  • Feature a sterilization indicator patch that darkens when exposed to steam sterilization for 20 minutes
  • Extra large sizes available, call for quote
Specifications

Specifications

Yes
Biohazard Bag
Yes
HDPE (High-density Polyethylene)
960mm
50μm
Red
790mm
Videos