missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Polypropylene Analytical Funnels

Manufactured with polypropylene for strength and durability. Kartell™ Polypropylene Analytical Funnels are designed specifically for analytical chemistry.

Brand:  Kartell™ 0015000

Code : A0

Additional Details : Weight : 0.30600kg

Product Code. 12523197

  • £189.00 / Pack of 20
Estimated Shipment: 09-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Manufactured with polypropylene for strength and durability. Kartell™ Polypropylene Analytical Funnels are designed specifically for analytical chemistry.

  • Takes standard size filter paper
  • 60° angled sides with internal and external ribs to speed filtration and avoid pressure lock
  • Withstands temperatures up to 135°C for short periods
  • Autoclavable at 121°C for 20min
  • Suitable for foodstuff
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers