missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HDAC3 Antibody (PCRP-HDAC3-2D4), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-14060MFV450
This item is not returnable.
View return policy
Description
HDAC3 Monoclonal antibody specifically detects HDAC3 in Human samples. It is validated for Flow Cytometry
Specifications
| HDAC3 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| PCRP-HDAC3-2D4 | |
| Flow Cytometry | |
| EC 3.5.1.98, HD3RPD3-2RPD3, histone deacetylase 3, SMAP45 | |
| Recombinant full-length human HDAC3 protein (Uniprot: O15379) | |
| 0.1 mL | |
| Apoptosis, Breast Cancer, Cancer, Chromatin Research, Epigenetics | |
| 8841 | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction