missing translation for 'onlineSavingsMsg'
Learn More

HC 067047, Tocris Bioscience™

Potent and selective TRPV4 antagonist. Restrictions apply. See special conditions on product page.

Brand:  Tocris (Bio-Techne) 410010

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13464144

  • £145.80 / 10mg


Estimated Shipment
03-04-2019
Add to basket

Description

Description

Potent and selective TRPV4 antagonist. Reversibly inhibits currents through mouse, human and rat TRPV4 orthologs (IC 50 values are 17, 48 and 133 nM respectively). Also inhibits the endogenous TRPV4-mediated response to 4α-PDH (IC 50 = 22 nM). Selective for TRPV4 over TRPV1, TRPV2, TRPV3 and TRPM8 channels.
Specifications

Chemical Identifiers

883031-03-6
471.524
2-methyl-1-3-4-morpholinyl propyl-5-phenyl-n-3-trifluoromethyl phenyl-1h-pyrrole-3-carboxamide, d0b1qm, 2-methyl-1-3-morpholin-4-ylpropyl-5-phenyl-n-3-trifluoromethyl phenyl pyrrole-3-carboxamide, hc hplc, 2-methyl-1-3-morpholin-4-yl propyl-5-phenyl-n-3-trifluoromethyl phenyl-1h-pyrrole-3-carboxamide, n-3-trifluoromethyl phenyl-2-methyl-1-3-morpholinopropyl-5-phenyl-1h-pyrrole-3-carboxamide
2-methyl-1-(3-morpholin-4-ylpropyl)-5-phenyl-N-[3-(trifluoromethyl)phenyl]pyrrole-3-carboxamide
C26H28F3N3O2
NCZYSQOTAYFTNM-UHFFFAOYSA-N
2742550
CC1=C(C=C(N1CCCN2CCOCC2)C3=CC=CC=C3)C(=O)NC4=CC=CC(=C4)C(F)(F)F

Specifications

HC 067047
883031-03-6
2-methyl-1-3-4-morpholinyl propyl-5-phenyl-n-3-trifluoromethyl phenyl-1h-pyrrole-3-carboxamide, d0b1qm, 2-methyl-1-3-morpholin-4-ylpropyl-5-phenyl-n-3-trifluoromethyl phenyl pyrrole-3-carboxamide, hc hplc, 2-methyl-1-3-morpholin-4-yl propyl-5-phenyl-n-3-trifluoromethyl phenyl-1h-pyrrole-3-carboxamide, n-3-trifluoromethyl phenyl-2-methyl-1-3-morpholinopropyl-5-phenyl-1h-pyrrole-3-carboxamide
CC1=C(C=C(N1CCCN2CCOCC2)C3=CC=CC=C3)C(=O)NC4=CC=CC(=C4)C(F)(F)F
471.524
471.51
10mg
C26H28F3N3O2
NCZYSQOTAYFTNM-UHFFFAOYSA-N
2-methyl-1-(3-morpholin-4-ylpropyl)-5-phenyl-N-[3-(trifluoromethyl)phenyl]pyrrole-3-carboxamide
2742550
>99%
Videos