missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ HB-2-30
Click to view available options
:
100 μg
Description
Key information: HB-2-30 is a far-red fluorescent transglutaminase-2 (TG2) probe for imaging endocytosis in vitro and in vivo. HB-2-30 undergoes highly efficient endocytosis in complex with TG2 and α2-macroglobulin via the LRP1 pathway.
Used for: Imaging endocytosis in vitro and in vivo, explore targeted lysosomal degradation of secreted and cell surface proteins.
Application: Confocal microscopy.
Properties and Photophysical Data: Suitable for multiplexing and can be combined with DAPI (Cat. No. 5748) counter-staining. Excitation and emission maxima (λ) are 649 and 665 nm, respectively.
Specifications
Specifications
| Purity | 90% |
| CAS | 3031522-14-9 |
| Content And Storage | Store at -20°C |
| Quantity | 100 μg |
| Target Entry | Fluorescent Lysosome Probes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction