missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Alfa Aesar Haynes™ 25 gauze, 60 mesh woven from 0.14mm (0.0055 in.) dia. wire

Brand:  Thermo Scientific Alfa Aesar 046866.GT

Haynes™ 25 gauze, 60 mesh woven from 0.14mm (0.0055 in.) dia. wire

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 15467335

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.