missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Haynes™ 25 gauze, 30 mesh woven from 0.28mm (0.011 in.) dia. wire

Brand:  Alfa Aesar™ 046892.FL

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 75 x 75mm

 View more versions of this product

Product Code. 15487335

  • £20.30 / Each


Estimated Shipment date
from Supplier 07-06-2018
Add to basket

Specifications

Chemical Identifiers

Haynes™ 25 gauze
30 mesh woven from 0.28mm (0.011 in.) dia. wire

Specifications

Haynes™ 25 gauze
75 x 75mm
Wire Cloth
30 Mesh
30 mesh woven from 0.28mm (0.011 in.) dia. wire
Videos
SDS