missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Haynes™ 25 gauze, 20 mesh woven from 0.13mm (0.005 in.) dia. wire

Brand:  Alfa Aesar™ 046645.HB

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 300 x 300mm

 View more versions of this product

Product Code. 15457275

  • £134.60 / Each


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

Haynes™ 25 gauze
20 mesh woven from 0.13mm (0.005 in.) dia. wire

Specifications

Haynes™ 25 gauze
300 x 300mm
Wire Cloth
20 Mesh
20 mesh woven from 0.13mm (0.005 in.) dia. wire
Videos
SDS