Learn More
Hastelloy C™ gauze, 40 mesh woven from 0.19mm (0.0075 in.) dia. wire, Thermo Scientific Chemicals
£25.60 - £280.00
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15427285
|
Thermo Scientific Alfa Aesar
046690.FL |
75 x 75 mm |
£25.60
Each |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15437285
|
Thermo Scientific Alfa Aesar
046690.GT |
150 x 150 mm |
£76.60
Each |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15447285
|
Thermo Scientific Alfa Aesar
046690.HB |
300 x 300 mm |
£280.00
Each |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Hastelloy C is used in combustion chambers, heat-treating equipment, collector rings, combustion chambers and exhaust stacks for high-heat applications in for conventional aircraft and jets, pipe connections, pumps and valves for oil refinery, chemical and petrochemical applications. Also used in components exposed to flue gas or flue gas desulfurization plants.