Hanging Wafer Robotic Tips: Tecan-Style Clear Tips, Presterilized

Clear tips for accurate sample delivery

Overview
Brand: Axygen

Manufacturer Part Number: TT-20-C-HTR-S

UNSPSC: 41121608

Code: 68

Additional Details:
Additional Details: Weight: 4.10000kg



Product Code. 11304295

 N/A
N/A

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description and Specification

Specification

Product Type Robotic Tip
Volume (Metric) 20μL
Sterility Sterile
Color Clear
Autoclavable Yes
Tip Style Racked
Shape Round
Material Polypropylene
Packaging Rack
Quantity 96 Tips/Rack
Certifications/Compliance Non-ANSI
For Use With (Application) For accurate sample delivery
For Use With (Equipment) Tecan™ Genesis™ Freedom™/Freedom Evo™/Miniprep with LiHa-1, 4, 8 head, Aurora Biomed Versa™ with 8-channel head